mFluor™ Violet 450-Wheat Germ Agglutinin (WGA) Conjugate
Katalogové číslo AATB25450
Dodavatel: AAT BIOQUEST
New Product
Číslo CAS:
Specifikace
- Bal.:1 mg
- Popis:mFluor™ Violet 450-Wheat germ agglutinin (WGA) conjugate
Specifikace
O tomto produktu
mFluor™ Violet 450 labelled WGA is well-excited by the violet laser, emitting a bright blue fluorescence at 445 nm.
- Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.