iFluor® 700-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # AATB25561
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Bal.:1 mg
- Popis:iFluor® 700-Wheat germ agglutinin (WGA) conjugate
Specifications
About this item
The iFluor® 700 conjugate of WGA possesses a bright, far-red fluorescence characteristic of the iFluor® 700 dye (Ex/Em = 690/713 nm).
- Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.