Orders LinkContactUsLinkComponent
FluoroQuest™ PLUS Antifade Mounting Medium
FluoroQuest™ PLUS Antifade Mounting Medium
FluoroQuest™ PLUS antifade mounting medium, 5 mL
Katalogové číslo AATB20008
Dodavatel:   AAT Bioquest
FluoroQuest™ PLUS Antifade Mounting Medium
Katalogové číslo AATB20008
Dodavatel:  AAT Bioquest
Číslo CAS:  
FluoroQuest™ PLUS Antifade Mounting Medium
FluoroQuest™ PLUS Antifade Mounting Medium
FluoroQuest™ PLUS antifade mounting medium, 5 mL
Přejít na:
Číslo CAS:  
Price
4 860,00 Kč
Přihlásit se pro dostupnost
1 * 5 mL
Product Details & Documents
FluoroQuest™ PLUS Antifade mounting medium, the latest breakthrough in non-setting media technology, provides unparalleled protection against fading throughout the visible spectrum, including far-red wavelengths.
  • Stable for 12 months
Quality tested in-house.
Upozornení: For research use only.
Specifikace
Specifikace
  • Bal.:
    5 mL
  • Popis:
    FluoroQuest™ PLUS antifade mounting medium