Order Entry
Czech Republic
ContactUsLinkComponent
Supelco® Organic Reference Standard, 1,3,5-Tri-tert-butylbenzene
Supelco® Organic Reference Standard, 1,3,5-Tri-tert-butylbenzene
Catalog # 94056-100MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, 1,3,5-Tri-tert-butylbenzene
Catalog # 94056-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Bal.:
    100 mg
  • Standard:
    Analytical standard
  • Čí. CAS:
    1460-02-2
  • Popis:
    1,3,5-Tri-tert-butylbenzene
  • Podmínky uskladnění:
    No temperature limit
  • Podmínky k odeslání:
    STD
  • MDL čí.:
    MFCD00008831
  • M-Clarity quality level:
    MQ100
  • Bod tání:
    67 - 72 °C (lit.)
  • Vzorec:
    C₆H₃[C(CH₃)₃]₃
  • Molekulární hmotnost:
    246,43
  • Bod varu:
    121 - 122 °C/12 mmHg (lit.)

Specifications

About this item

The 135-Tri-tert-butylbenzene may find the following uses:

  • The qualitative study of pyrolytic product vapor of Helianthus annuus stems using pyrolysis–gas chromatography-mass spectrometry (Py-GC/MS)
  • Gas-phase synthesis and Fourier-transform ion cyclotron resonance (FT/ICR) mass spectrometric characterisation of sandwich complexes of Sc+, Y+, and lanthanide ions with 1,3,5 -tri-tert-butylbenzene