ROXtra SSL-NHS ester, ReadiCleave™
2070, 1 mg
Katalogové číslo AATB7052
Dodavatel: AAT Bioquest
Přejít na:
Product Details & Documents
ReadiCleave™ ROXtra succinimidyl ester contains a S-S linker that can be cleaved with TCEP or DTT to remove the ROXtra fluorophore from the target molecule.
- Stable for 12 months
AAT Bioquest’s ReadiCleave™ linkers enable fluorescent tags conjugated to a biological target from which the added fluorescent tag can be removed when needed.
Upozornení: For Research Use Only
- Molecular Weight:1193.49 g/mol
Specifikace
Specifikace
- Bal.:1 mg
- Sekvence:2070