Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
CDPI3-CPG
CDPI3-CPG (500 A) [Minor Groove Binder CPG]
CDPI3-CPG (500 A) [Minor Groove Binder CPG]
AATB6900
:
AAT Bioquest
New Product
:
CDPI3-CPG (500 A) [Minor Groove Binder CPG]
AATB6900
:
AAT Bioquest
New Product
:
Bal.:
100 mg
Sekvence:
2440
: