Anti-MEK1 + MEK2 Rabbit Polyclonal Antibody
ANTIA93672-100
New Product
- Druh protilátky:Primární
- Název antigenu:AA589381
- Symbol antigenu:MEK1 + MEK2
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# Q02750
- Synonyma antigenu:MAP kinase kinase 2|Dual specificity mitogen-activated protein kinase kinase 1|CFC3|ERK activator kinase 2|ERK activator kinase 1|EC 2.7.12.2|Dual specificity mitogen-activated protein kinase kinase 2|FLJ26075|MAP kinase kinase 1
- Skladovací pufr:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molekulární hmotnost:43 kDa
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at 4 °C
- Imunogen:Synthetic phospho-peptide derived from human MAP2K1 around the phosphorylation site of Thr386
- Čištění:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Typ balení:Plastic vial
- Bal.:100 µG
Rabbit polyclonal antibody to MAP2K1 (phospho Thr386) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: MEK1 + MEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat