Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CD99R Mouse Monoclonal Antibody [Clone: MEM-131] (PE (Phycoerythrin))
  ANTIA86614-100
New Product
Anti-CD99R Mouse Monoclonal Antibody [Clone: MEM-131] (PE (Phycoerythrin))
  ANTIA86614-100
New Product

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    1110061M03Rik
  • Symbol antigenu:
    CD99R
  • Klonovalita:
    Monoclonal
  • Klon:
    MEM-131
  • Spojení:
    PE (Phycoerythrin)
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Mouse
  • Izotyp:
    IgM
  • Reaktivita:
    Human
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    23449
  • Synonyma antigenu:
    CD99|12E7|CD99 molecule|2410026K10Rik|antigen identified by monoclonal antibodies 12E7, F21 and O13|CD99 antigen|antigen identified by monoclonal 12E7, Y homolog|CD99R|CD99 antigen, X chromosome
  • Skladovací pufr:
    Supplied in Tris Buffered Saline with 15 mM Sodium Azide
  • Skladovací teplota:
    Store in the dark at 2...8 °C. Do not freeze!
  • Transportní teplota:
    Shipped on blue ice at 4 °C
  • Imunogen:
    HPB-ALL human peripheral blood leukemia T-cell line
  • Čištění:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Typ balení:
    Glass bottle
  • Bal.:
    100 T

 

 

Mouse monoclonal [MEM-131] antibody to CD99R (PE) for Flow Cytometry with samples derived from Human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD99R
Clonality: Monoclonal
Clone: MEM-131
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human