Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MEK1 + MEK2 Rabbit Polyclonal Antibody
Anti-MEK1 + MEK2 Rabbit Polyclonal Antibody
Catalog # ANTIA95531-100
Supplier:  Antibodies.com
New Product
undefined
Anti-MEK1 + MEK2 Rabbit Polyclonal Antibody
Catalog # ANTIA95531-100
Supplier:  Antibodies.com
New Product

Specifications

  • Druh protilátky:
    Primární
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    UniprotID# Q02750, P36507
  • Skladovací pufr:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molekulární hmotnost:
    43 kDa
  • Skladovací teplota:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Transportní teplota:
    Shipped on blue ice at +4 °C
  • Imunogen:
    Synthetic peptide derived from human MEK1/2 (amino acids 189-238)
  • Čištění:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Typ balení:
    Plastic vial
  • Bal.:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to MEK1 + MEK2 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat