Anti-Dynamin 1 + 2 Rabbit Polyclonal Antibody
Catalog # ANTIA95185-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Druh protilátky:Primární
- Název antigenu:CMTDI 1
- Symbol antigenu:Dynamin 1 + 2
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# Q05193
- Synonyma antigenu:CMTDIB|DNM1|DNM2|CMTDI1|DNM 1|Dynamin|DYN2|DNM 2|DYN II
- Skladovací pufr:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molekulární hmotnost:97 kDa
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at +4 °C
- Imunogen:Synthetic peptide derived from human Dynamin-1 (amino acids 740-789)
- Testované aplikace:IHC
- Čištění:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Typ balení:Plastic vial
- Bal.:100 µG
Specifications
About this item
Rabbit polyclonal antibody to Dynamin-1 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:5000
Type: Primary
Antigen: Dynamin 1 + 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat