Anti-Integrin beta 1d Rabbit Polyclonal Antibody
Katalogové číslo ANTIA95152-100
Dodavatel: ANTIBODIES.COM
New Product
Specifikace
- Druh protilátky:Primární
- Název antigenu:CD29
- Symbol antigenu:Integrin beta 1d
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# P05556
- Synonyma antigenu:FNRB|Integrin beta 1|VLAB|MDF2|ITGB 1|MSK12|Integrin beta 1d|ITGB1
- Skladovací pufr:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molekulární hmotnost:88 kDa
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at +4 °C
- Imunogen:Synthetic peptide derived from human Integrin beta1 (amino acids 733-782)
- Čištění:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Typ balení:Plastic vial
- Bal.:100 µG
Specifikace
O tomto produktu
Rabbit polyclonal antibody to Integrin beta1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Integrin beta 1d
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat