Anti-Brk Rabbit Polyclonal Antibody
Katalogové číslo ANTIA95137-100
Dodavatel: ANTIBODIES.COM
New Product
Specifikace
- Druh protilátky:Primární
- Název antigenu:Breast tumor kinase
- Symbol antigenu:Brk
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Mouse
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# Q13882
- Synonyma antigenu:Tyrosine protein kinase BRK|Tyrosine-protein kinase BRK|Breast tumour kinase|Protein-tyrosine kinase 6|Protein tyrosine kinase 6|PTK6_HUMAN|Brk|Ptk6
- Skladovací pufr:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molekulární hmotnost:51 kDa
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at +4 °C
- Imunogen:Synthetic peptide derived from human Breast Tumor Kinase (amino acids 10-59)
- Čištění:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Typ balení:Plastic vial
- Bal.:100 µG
Specifikace
O tomto produktu
Rabbit polyclonal antibody to Breast Tumor Kinase for WB and ELISA with samples derived from Human and Mouse.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: Brk
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse