Anti-MEK1 (phospho Ser298) Rabbit Monoclonal Antibody [clone: ARC1565]
Catalog # ANTIA308195-100
Supplier: Antibodies.com
New Product
Specifications
- Druh protilátky:Primární
- Název antigenu:Dual specificity mitogen activated protein kinase kinase 1
- Symbol antigenu:MEK1
- Klonovalita:Monoclonal
- Klon:ARC1565
- Spojení:Unconjugated
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# Q02750
- Synonyma antigenu:MAPKK 1|Dual specificity mitogen-activated protein kinase kinase 1|MAP2K1|MAPKK1|MAP kinase/Erk kinase 1|MEK 1|ERK activator kinase 1|MAP kinase kinase 1|MAPK/ERK kinase 1
- Modifikace:phospho Ser298
- Skladovací pufr:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molekulární hmotnost:45 kDa
- Sekvence:PLSSY
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at 4 °C
- Imunogen:A synthetic phosphorylated peptide around S298 of human Phospho-MEK1-S298 (Q02750)
- Čištění:Affinity purification
- Typ balení:Plastic vial
- Bal.:100 µl
Specifications
About this item
Rabbit monoclonal [ARC1565] antibody to MEK1 (phospho Ser298) for WB with samples derived from human and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: MEK1
Clonality: Monoclonal
Clone: ARC1565
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat