Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IRE1 Rabbit Polyclonal Antibody
  ANTIA98163-100
New Product
undefined
Anti-IRE1 Rabbit Polyclonal Antibody
  ANTIA98163-100
New Product

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Endoplasmic reticulum (ER) to nucleus signalling 1
  • Symbol antigenu:
    IRE1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Mouse
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    UniprotID# Q76MJ5
  • Synonyma antigenu:
    ERN1_HUMAN|Endoribonuclease|Endoplasmic reticulum-to-nucleus signaling 1|hIRE1p|ERN 1|Endoplasmic reticulum to nucleus signaling 1|hIRE 1p|ER to nucleus signaling 1|Ern1
  • Skladovací pufr:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molekulární hmotnost:
    102 kDa
  • Skladovací teplota:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Transportní teplota:
    Shipped on blue ice at +4 °C
  • Imunogen:
    Synthetic peptide derived from human ERN2 (amino acids 841-890)
  • Testované aplikace:
    IHC
  • Čištění:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Typ balení:
    Plastic vial
  • Bal.:
    100 µG

 

 

Rabbit polyclonal antibody to ERN2 for IHC, IF and ELISA with samples derived from Human and Mouse.

  • Validated Applications: IHC, IF, ELISA
  • Recommended Dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:10000

Type: Primary
Antigen: IRE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse