Order Entry
Czech Republic
ContactUsLinkComponent
Anti-POU2F1 Goat Polyclonal Antibody
Anti-POU2F1 Goat Polyclonal Antibody
Catalog # ANTIA84809-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-POU2F1 Goat Polyclonal Antibody
Catalog # ANTIA84809-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    FLJ42836
  • Symbol antigenu:
    POU2F1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Goat
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Formulář:
    Liquid
  • Synonyma antigenu:
    Octamer binding transcription factor 1|NF A1|Octamer-binding protein 1|45200|Oct 1B|NF-A1|Octamer binding protein 1
  • Skladovací pufr:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sekvence:
    C-NVQSKSNEESGDSQQ
  • Skladovací teplota:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Koncentrace:
    100 µg at 0,5 mg/ml
  • Transportní teplota:
    Shipped on blue ice at 4 °C
  • Imunogen:
    Synthetic peptide corresponding to Human POU2F1 (internal region)
  • Čištění:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Typ balení:
    Plastic vial
  • Bal.:
    100 µG

Specifications

About this item

Goat polyclonal antibody to POU2F1 for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, W

Type: Primary
Antigen: POU2F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human