Anti-EGF Mouse Monoclonal Antibody [clone: KT57]
Dodavatel: ANTIBODIES.COM
New Product
Certifikáty
O tomto produktu
Mouse monoclonal (KT57) antibody to EGF for ELISA, WB and sELISA with samples derived from human.
- Validated applications: ELISA, WB, sELISA
- Recommended dilutions: ELISA: 1 µg/ml, WB: 1/500 to 1/5000, sELISA: 0,5 to 2 µg/ml
Type: Primary
Antigen: EGF
Clonality: Monoclonal
Clone: KT57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifikace
- Catalog No:
- ANTIA60-200
- ANTIA60-100
- Symbol antigenu:
- EGF
- EGF
- Název antigenu:
- AI790464
- AI790464
- Synonyma antigenu:
- OTTHUMP00000219722|OTTHUMP00000219721|EGF|beta-urogastrone|EGF_HUMAN|Epidermal growth factor|Pro epidermal growth factor|HOMG4|Beta urogastrone
- OTTHUMP00000219722|OTTHUMP00000219721|EGF|beta-urogastrone|EGF_HUMAN|Epidermal growth factor|Pro epidermal growth factor|HOMG4|Beta urogastrone
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Monoclonal
- Monoclonal
- Spojení:
- Unconjugated
- Unconjugated
- Klon:
- KT57
- KT57
- Reaktivita:
- Human
- Human
- Hostitel:
- Mouse
- Mouse
- Identifikační číslo genu:
- UniprotID# P01133
- UniprotID# P01133
- Izotyp:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Imunogen:
- Recombinant human EGF
- Recombinant human EGF
- Formulář:
- Liquid
- Liquid
- Čištění:
- IgG is purified through a Protein A column.
- IgG is purified through a Protein A column.
- Koncentrace:
- 1 mg/ml
- 1 mg/ml
- Skladovací pufr:
- Supplied in phosphate buffered saline with 0,09% sodium azide
- Supplied in phosphate buffered saline with 0,09% sodium azide
- Skladovací teplota:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Testované aplikace:
- Sandwich ELISA
- Sandwich ELISA
Specifikace
Frequently Bought Together