Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Telethonin Rabbit Polyclonal Antibody
  ANTIA9992-100
New Product
undefined
Anti-Telethonin Rabbit Polyclonal Antibody
  ANTIA9992-100
New Product

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    19 kDa sarcomeric protein
  • Symbol antigenu:
    Telethonin
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    UniprotID# O15273
  • Synonyma antigenu:
    LGMD2G|TELE|T-cap|Limb girdle muscular dystrophy 2G (autosomal recessive)|CMD1N|Tcap|CMH25|TELT_HUMAN|Telethonin
  • Skladovací pufr:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molekulární hmotnost:
    21/19 kDa
  • Sekvence:
    MATSELSCEVSEENCERREAFWAEWKDLTLSTRPEEGCSLHEEDTQRHETYHQQGQCQVLVQRSPWLMMRMGILGRGLQEYQLPYQRVLPLPIFTPAKMGATKEEREDTPIQLQELLALETALGGQCVDRQEVAEITKQLPPVVPVSKPGALRRSLSRSMSQEAQRG
  • Skladovací teplota:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Transportní teplota:
    Shipped on blue ice at 4 °C
  • Imunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-167 of human TCAP (NP_003664.1)
  • Čištění:
    Affinity purification
  • Typ balení:
    Plastic vial
  • Bal.:
    100 µl

 

 

Rabbit polyclonal antibody to Telethonin for WB with samples derived from mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:100 to 1:50

Type: Primary
Antigen: Telethonin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat