Anti-TOR1AIP1 Rabbit Polyclonal Antibody
Catalog # ANTIA99939-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Druh protilátky:Primární
- Název antigenu:Lamin-associated protein 1B
- Symbol antigenu:TOR1AIP1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# Q5JTV8
- Synonyma antigenu:Lamina associated polypeptide 1C|Lamina associated polypeptide 1B|TOR1AIP1|LAP1B|Torsin-1A-interacting protein 1|TOIP1_HUMAN|Torsin A interacting protein 1|LAP1
- Skladovací pufr:Supplied in Phosphate Buffered Saline (without Mg⁺² and Ca⁺²), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molekulární hmotnost:66 kDa
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at 4 °C
- Imunogen:Synthetic peptide derived from human TOR1AIP1 (amino acids 431-480)
- Čištění:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Typ balení:Plastic vial
- Bal.:100 µG
Specifications
About this item
Rabbit polyclonal antibody to TOR1AIP1 for WB and ELISA with samples derived from human.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: TOR1AIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human