Anti-ASK1 Rabbit Polyclonal Antibody
Catalog # ANTIA99927-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Druh protilátky:Primární
- Název antigenu:Apoptosis signal regulating kinase 1
- Symbol antigenu:ASK1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# Q99683
- Synonyma antigenu:ASK 1|ASK-1|MAPK/ERK kinase kinase 5|MAP3K5|MAP/ERK kinase kinase 5|Apoptosis signal-regulating kinase 1|M3K5_HUMAN|ASK1|M3K5
- Skladovací pufr:Supplied in Phosphate Buffered Saline (without Mg⁺² and Ca⁺²), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molekulární hmotnost:154 kDa
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at 4 °C
- Imunogen:Synthetic peptide derived from human ASK1 (amino acids 49-98)
- Testované aplikace:IHC
- Čištění:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Typ balení:Plastic vial
- Bal.:100 µG
Specifications
About this item
Rabbit polyclonal antibody to ASK1 for WB, IHC and ELISA with samples derived from human.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:20000
Type: Primary
Antigen: ASK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human