Anti-SERPINB4 Rabbit Polyclonal Antibody
Catalog # ANTIA99944-100
Dodavatel: ANTIBODIES.COM
New Product
Specifikace
- Druh protilátky:Primární
- Název antigenu:Leupin
- Symbol antigenu:SERPINB4
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# P48594
- Synonyma antigenu:SCCA-2|PI11|SCCA 2|Peptidase inhibitor 11|Protease inhibitor (leucine serpin)|SCCA2|SCCA1|PI 11|PI-11
- Skladovací pufr:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molekulární hmotnost:44 kDa
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at 4 °C
- Imunogen:Synthetic peptide derived from human SERPINB4 (amino acids 261-310)
- Čištění:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Typ balení:Plastic vial
- Bal.:100 µG
Specifikace
O tomto produktu
Rabbit polyclonal antibody to SERPINB4 for WB and ELISA with samples derived from human.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: SERPINB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human