Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PSD93 Rabbit Polyclonal Antibody
Anti-PSD93 Rabbit Polyclonal Antibody
Catalog # ANTIA99910-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-PSD93 Rabbit Polyclonal Antibody
Catalog # ANTIA99910-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Channel associated protein of synapse 110
  • Symbol antigenu:
    PSD93
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Formulář:
    Liquid
  • Identifikační číslo genu:
    UniprotID# Q15700
  • Synonyma antigenu:
    Channel-associated protein of synapse-110|DKFZp781E0954|Discs, large homolog 2 (Drosophila)|Channel associated protein of synapses 110kD|Chapsyn110|Chapsyn-110|DKFZp781D1854|Disks large homolog 2|Chapsyn 110
  • Skladovací pufr:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molekulární hmotnost:
    97 kDa
  • Skladovací teplota:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Transportní teplota:
    Shipped on blue ice at 4 °C
  • Imunogen:
    Synthetic peptide derived from human DLG2 (amino acids 601-650)
  • Testované aplikace:
    IHC
  • Čištění:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Typ balení:
    Plastic vial
  • Bal.:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to DLG2 for WB, IHC and ELISA with samples derived from human.

  • Validated applications: WB, IHC, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen: PSD93
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human