Anti-UGT2B17 Rabbit Polyclonal Antibody
Katalogové číslo ANTIA306025-100
Dodavatel: ANTIBODIES.COM
New Product
Specifikace
- Druh protilátky:Primární
- Symbol antigenu:UGT2B17
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Human
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# O75795
- Skladovací pufr:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Sekvence:GHEVIVLTSSASILVNASKSSAIKLEVYPTSLTKNDLEDFFMKMFDRWTYSISKNTFWSYFSQLQELCWEYSDYNIKLCEDAVLNKKLMRKLQESKFDVLLADAVNPCGELLAELLNIPFLYSLRFSVGYTVEKNGGGFLFPPSYVPVVMS
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at 4 °C
- Imunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 50-200 of human UGT2B17 (NP_001068.1).
- Testované aplikace:ICC/IF
- Čištění:Affinity purification
- Typ balení:Plastic vial
- Bal.:100 µl
Specifikace
O tomto produktu
Rabbit polyclonal antibody to UGT2B17 for ICC/IF with samples derived from Human.
- Validated Applications: ICC/IF
- Recommended Dilutions: ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: UGT2B17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human