Anti-MEK1 Rabbit Monoclonal Antibody [clone: ARC0044]
Katalogové číslo ANTIA307613-100
Dodavatel: Antibodies.com
New Product
Specifikace
- Druh protilátky:Primární
- Název antigenu:Dual specificity mitogen activated protein kinase kinase 1
- Symbol antigenu:MEK1
- Klonovalita:Monoclonal
- Klon:ARC0044
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunochemie:Yes
- Imunoprecipitace:Yes
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# Q02750
- Synonyma antigenu:MAPKK 1|Dual specificity mitogen-activated protein kinase kinase 1|MAP2K1|MAPKK1|MAP kinase/Erk kinase 1|MEK 1|ERK activator kinase 1|MAP kinase kinase 1|MAPK/ERK kinase 1
- Skladovací pufr:Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
- Molekulární hmotnost:45 kDa
- Sekvence:MPKKKPTPIQLNPAPDGSAVNGTSSAETNLEALQKKLEELELDEQQRKRLEAFLTQKQKVGELKDDDFEKISELGAGNGGVVFKVSHKPSGLVMARKLIH
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at 4 °C
- Imunogen:A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEK1 (Q02750)
- Testované aplikace:ICC/IF
- Čištění:Affinity purification
- Typ balení:Plastic vial
- Bal.:100 µl
Specifikace
O tomto produktu
Rabbit monoclonal [ARC0044] antibody to MEK1 for WB, ICC/IF and IP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ICC/IF, IP
- Recommended Dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:200, IP: 1:500 to 1:1000
Type: Primary
Antigen: MEK1
Clonality: Monoclonal
Clone: ARC0044
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat