ReadiUse™ dNTP Mix Set *10 mM PCR Grade*
Catalog # AATB17258
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Bal.:1 mL
Specifications
About this item
10 mM dNTP Mix is a mixture of four nucleotides (dATP, dCTP, dGTP, dTTP) in purified water.
- Each nucleotide is at a concentration of 10 mM
- The formulation is stable for 12 months at −20 °C
10 mM dNTP mix is suitable for use in polymerase chain reaction (PCR), sequencing, fill-in reactions, nick translation, cDNA synthesis, and TdT-tailing reactions.
Caution: For research use only.