Fluorescein-12-dCTP *1 mM in Tris Buffer (pH 7,5)*
Catalog # AATB17035
Supplier: AAT Bioquest
New Product
CAS Number:
Specifications
- Bal.:25 Nmo
Specifications
About this item
Fluorescein-12-dCTP might be incorporated into a DNA sequence as its natural counterpart dCTP to incorporate a fluorescein tag in a DNA molecule.
- Can be used for the direct enzymatic labelling of DNA/cDNA, e.g., by PCR or nick translation
The linker between fluorescein tag and cytidine has been optimised for the optimal substrate incorporation and labelling efficiency.
Caution: For research use only.