Anti-HIF-1 alpha Goat Polyclonal Antibody
ANTIA121625-600
New Product
- Druh protilátky:Primární
- Název antigenu:ARNT interacting protein
- Symbol antigenu:HIF-1 alpha
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Goat
- Izotyp:IgG
- Reaktivita:Human,Dog,Mouse
- Western blot:Yes
- Formulář:Liquid
- Synonyma antigenu:HIF 1alpha|ARNT-interacting protein|Basic-helix-loop-helix-PAS protein MOP1|HIF-1-alpha|HIF-1 alpha|Basic helix loop helix PAS protein MOP1|bHLHe78|HIF 1A|Class E basic helix-loop-helix protein 78
- Skladovací pufr:Supplied in Phosphate Buffered Saline with 20% Glycerol and 0,05% Sodium Azide
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
- Koncentrace:3 mg/ml
- Transportní teplota:Shipped on blue ice at 4 °C
- Imunogen:Recombinant peptide derived from within residues a.a. 780 to the C terminus of human HIF-1 alpha, expressed in and purified from E. coli
- Čištění:Affinity purification
- Typ balení:Plastic vial
- Bal.:600 µG
Goat polyclonal antibody to HIF-1 alpha for WB with samples derived from human, mouse and canine.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: HIF-1 alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Dog