Anti-T7 Tag Goat Polyclonal Antibody
Katalogové číslo ANTIA121634-300
Dodavatel: ANTIBODIES.COM
New Product
Specifikace
- Druh protilátky:Primární
- Symbol antigenu:T7 Tag
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Goat
- Izotyp:IgG
- Western blot:Yes
- Formulář:Liquid
- Skladovací pufr:Supplied in Phosphate Buffered Saline with 20% Glycerol and 0,05% Sodium Azide
- Sekvence:MASMTGGQQMG
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
- Koncentrace:3 mg/ml
- Transportní teplota:Shipped on blue ice at 4 °C
- Imunogen:Recombinant T7 Tag peptide, expressed in and purified from E. coli
- Čištění:Affinity purification
- Typ balení:Plastic vial
- Bal.:300 µG
Specifikace
O tomto produktu
Goat polyclonal antibody to T7 Tag for WB.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:5,000
- Host: Goat
- Clonality: Polyclonal
- Isotype: IgG
- Conjugate: Unconjugated
Type: Primary
Antigen: T7 Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: