Anti-REP1 Goat Polyclonal Antibody
New Product
Goat polyclonal antibody to REP1 for WB with samples derived from human, rat, mouse, canine and monkey.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: REP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat, Mouse, Dog, Monkey
- Catalog No:
- ANTIA121631-2
- ANTIA121631-800
- Symbol antigenu:
- REP1
- REP1
- Název antigenu:
- CHM
- CHM
- Synonyma antigenu:
- GGTA|DXS540|Rab geranylgeranyltransferase component A|Rab escort protein 1|FLJ38564|Choroideraemia protein|MGC102710|Choroideremia|HSD 32
- GGTA|DXS540|Rab geranylgeranyltransferase component A|Rab escort protein 1|FLJ38564|Choroideraemia protein|MGC102710|Choroideremia|HSD 32
- Druh protilátky:
- Primární
- Primární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- Unconjugated
- Unconjugated
- Reaktivita:
- Monkey, Human, Dog, Rat, Mouse
- Monkey, Human, Dog, Rat, Mouse
- Hostitel:
- Goat
- Goat
- Izotyp:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Imunogen:
- Recombinant peptide derived from within residues a.a. 617 to the C terminus of human REP1, expressed in and purified from E. coli
- Recombinant peptide derived from within residues a.a. 617 to the C terminus of human REP1, expressed in and purified from E. coli
- Formulář:
- Liquid
- Liquid
- Čištění:
- Affinity purification
- Affinity purification
- Koncentrace:
- 4 mg/ml
- 4 mg/ml
- Skladovací pufr:
- Supplied in Phosphate Buffered Saline with 20% Glycerol and 0,05% Sodium Azide
- Supplied in Phosphate Buffered Saline with 20% Glycerol and 0,05% Sodium Azide
- Skladovací teplota:
- Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
- Transportní teplota:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Frequently Bought Together