Fluorescein-12-dATP *1 mM in Tris Buffer (pH 7,5)*
Katalogové číslo AATB17036
Dodavatel: AAT Bioquest
New Product
Číslo CAS:
Specifikace
- Bal.:25 Nmo
Specifikace
O tomto produktu
Fluorescein-12-dATP might be incorporated into a DNA sequence as its natural counterpart dATP to incorporate a fluorescein tag in a DNA molecule.
- Can be used for the direct enzymatic labelling of DNA/cDNA, e.g., by PCR or Nick Translation
The linker between fluorescein tag and adenine has been optimised for the optimal substrate incorporation and labelling efficiency.
Caution: For research use only.