DEAC-dUTP *1 mM in TE Buffer (pH 7,5)*
AATB17034
New Product
:
- Bal.:100 Nmo
The dye-modified deoxyuridine 5'-triphosphates (such as aminoallyl-dUTP) can be used to produce dye-containing DNA by conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling or PCR.
- Chemically identical to Diethylaminocoumarin-5-dUTP (Perkin Elmer’s NEL455001EA)
This Coumarin-dUTP conjugate can be used as a blue fluorescence colour with Spectrum Aqua™ filter set (Spectrum Aqua™ is the trademark of Vysis).
: For research use only.