Anti-Rat IgG Fc Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Katalogové číslo ANTIA294684-500
Dodavatel: ANTIBODIES.COM
New Product
Specifikace
- Druh protilátky:Sekundární
- Symbol antigenu:Rat IgG Fc
- Klonovalita:Polyclonal
- Spojení:AP (Alkaline Phosphatase)
- ELISA:Yes
- Hostitel:Goat
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Rat
- Western blot:Yes
- Formulář:Liquid
- Fragment protilátky:F(ab')2
- Skladovací pufr:Supplied in 50 mM HEPES, pH 7,1, with 0,1 M sodium chloride, 1 mM magnesium chloride, 0,1 mM zinc chloride, 0,2% BSA, and 0,09% sodium azide
- Skladovací teplota:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze.
- Koncentrace:0,5 mg/ml
- Transportní teplota:Shipped on blue ice at 4 °C
- Testované aplikace:IHC, ICC
- Čištění:Antigen affinity purification
- Typ balení:Plastic vial
- Bal.:500 µG
Specifikace
O tomto produktu
Goat F(ab')2 anti-rat IgG Fc (AP) secondary antibody for WB, IHC, ICC and ELISA with samples derived from rat.
- Validated applications: WB, IHC, ICC, ELISA
- Recommended dilutions: ELISA: 1:2500 to 1:25000, ICC: 1:100 to 1:1000, IHC: 1:100 to 1:1000, WB: 1:2500 to 1:25000
Type: Secondary
Antigen: Rat IgG Fc
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat