Synthetic Maleimide Activated mKLH (from Megathura crenulata)
Katalogové číslo AATB5612
Dodavatel: AAT Bioquest
New Product
Číslo CAS:
Specifikace
- Typ balení:Vial
- Bal.:3
- Spojení:Unconjugated
- Typ proteinu/peptidu:Synthetic
- Zdroj:Megathura crenulata
- Podmínky uskladnění:Freeze (<−15 °C)
- Proteinová synonyma:Mariculture keyhole limpet hemocyanin
- Název proteinu/peptidu:Maleimide Activated mKLH
- Transportní teplota:RT
- Pk Info:3×2 mg
Specifikace
O tomto produktu
Maleimide Activated mKLH is engineered to facilitate the conjugation of sulfhydryl-containing peptide haptens to the highly immunogenic keyhole limpet hemocyanin (KLH) carrier protein, optimising it for immunisation and antibody production.
- Designed for heightened immunogen response
This high-purity, mariculture-derived KLH is chemically modified with a Sulfo-SMCC crosslinker, rendering it reactive to sulfhydryl groups (-SH) on cysteine residues of peptides and other thiol-containing haptens.
Caution: For research use only.