Mouse Immunoglobulin Isotyping ELISA Kit
Catalog # ANTIA246961-96
Supplier: ANTIBODIES.COM
New Product
CAS Number:
Specifications
- Bal.:96 T
- Doba trvání testu:Multiple steps
- Typ testu:Sandwich
- Formát:Pre-coated
- Primary antibody reactivity:Mouse
- Cíl:Immunoglobulin Isotyping
- Popis:Mouse immunoglobulin isotyping ELISA kit
- Typ vzorku:Serum, plasma, tissue homogenates, and other biological fluids
- Detekční metoda:Colorimetric
- Doba do výsledků:4 h 30 min
- Životnost police:Store for 6 months at 4 °C
- Skladovací teplota:4 °C
- Regulační status:RUO
Specifications
About this item
Mouse Immunoglobulin Isotyping ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qualitative determination of mouse Immunoglobulin Isotyping in serum, plasma, tissue homogenates, and other biological fluids.