Specifikace
- Bal.:96 Tests
- Doba trvání testu:One step
- Typ testu:Sandwich
- Formát:Pre-coated
- Primary antibody reactivity:Mouse
- Cíl:TGF beta Receptor III/TGFBR3
- Popis:Mouse TGF beta receptor III/TGFBR3 ELISA kit
- Typ vzorku:Serum, plasma or other biological fluids
- Detekční metoda:Colorimetric
- Doba do výsledků:1 h 30 min
- Životnost police:Store for 6 months at 4 °C
- Detekční rozsah:40 - 640 pg/ml
- Skladovací teplota:4 °C
- Objem vzorku:40 μl
- Citlivost:2,51 pg/ml
- Regulační status:RUO
Specifikace
O tomto produktu
Mouse TGF beta Receptor III/TGFBR3 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse TGF beta Receptor III/TGFBR3 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 40 to 640 pg/ml
- Sensitivity: 2,51 pg/ml
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse TGF beta Receptor III/TGFBR3 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse TGF beta Receptor III/TGFBR3 in An antibody specific for TGF beta Receptor III/TGFBR3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TGF beta Receptor III/TGFBR3 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-TGF beta Receptor III/TGFBR3 antibody, which also binds the TGF beta Receptor III/TGFBR3 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-TGF beta Receptor III/TGFBR3 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-TGF beta Receptor III/TGFBR3 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of TGF beta Receptor III/TGFBR3 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of TGF beta Receptor III/TGFBR3 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.