Human iNOS ELISA Kit
Product Details & Documents
Human iNOS ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human iNOS in serum, plasma, tissue homogenates, and other biological fluids.
- Ready-to-use ELISA kit
- Detection Range: 78,125 to 5000 pg/ml
- Sensitivity: 46,875 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%
Human iNOS ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human iNOS in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for iNOS has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the iNOS present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-iNOS Antibody, which binds the captured iNOS present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of iNOS captured in each well. The concentration of iNOS can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Specifikace
Specifikace
- Bal.:96 Tests
- Popis:Human iNOS ELISA kit
- Cíl:iNOS
- Primary antibody reactivity:Human
- Citlivost:46,875 pg/ml
- Typ vzorku:Serum, plasma, tissue homogenates and other biological fluids
- Princip testu:Quantitative
- Typ testu:Sandwich
- Objem vzorku:100 μl
- Detekční rozsah:78,125 - 5000 pg/ml
- Cat. No.:ANTIA74232-96
- Doba trvání testu:Multiple steps
- Detekční metoda:Colorimetric
- Doba do výsledků:4 h 30 min
- Formát:Pre-coated
- Skladovací teplota:4 °C
- Životnost police:Store for 6 months at 4 °C
- Křížová reakce:Human iNOS ELISA kit exhibits high specificity and excellent specificity for the detection of human iNOS. No significant cross-reactivity or interference between iNOS and analogues was observed.
- Regulační status:RUO



