Order Entry
Czech Republic
ContactUsLinkComponent
Mouse RELM beta ELISA Kit
Mouse RELM beta ELISA Kit
  ANTIA313993-96
New Product
 :  
undefined
Mouse RELM beta ELISA Kit
  ANTIA313993-96
New Product
 :  

 

  • Bal.:
    96 Tests
  • Doba trvání testu:
    One step
  • Typ testu:
    Sandwich
  • Formát:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Cíl:
    RELM beta
  • Popis:
    Mouse RELM beta ELISA kit
  • Typ vzorku:
    Serum, plasma or other biological fluids
  • Detekční metoda:
    Colorimetric
  • Doba do výsledků:
    1 h 30 min
  • Životnost police:
    Store for 6 months at 4 °C
  • Detekční rozsah:
    200 - 3200 ng/L
  • Skladovací teplota:
    4 °C
  • Objem vzorku:
    40 μl
  • Citlivost:
    9,84 ng/L
  • Regulační status:
    RUO

 

 

Mouse RELM beta ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse RELM beta in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 200 to 3200 ng/L
  • Sensitivity: 9,84 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse RELM beta ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse RELM beta in serum, plasma or other biological fluids. An antibody specific for RELM beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the RELM beta present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-RELM beta antibody, which also binds the RELM beta present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-RELM beta antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-RELM beta antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of RELM beta captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of RELM beta in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.