Cy5 Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Catalog # AATB67016
Supplier: AAT Bioquest
New Product
Specifications
- Frame colour:Bílá
- Poč. jamek:96
- Profil:Low
- Lem:Semi-skirted
- Well colour:Bílá
- Popis:Cy5 Dye qPCR calibration plate
- Čárový kód:Ano
- Bal.:1
Specifications
About this item
A Cy5 dye qPCR calibration plate is a type of plate used in quantitative PCR (polymerase chain reaction) experiments.
- The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
- This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimise light exposure.