Specifications
- Bal.:96 Tests
- Doba trvání testu:Multiple steps
- Typ testu:Sandwich
- Formát:Pre-coated
- Primary antibody reactivity:Human
- Cíl:TGF beta Receptor I
- Popis:Human TGF beta receptor I ELISA kit
- Typ vzorku:Serum, plasma, tissue homogenates and other biological fluids
- Křížová reakce:Human TGF beta Receptor I ELISA kit exhibits high specificity and excellent specificity for the detection of human TGF beta Receptor I. No significant cross-reactivity or interference between TGF beta Receptor I and analogues was observed.
- Detekční metoda:Colorimetric
- Doba do výsledků:4 h 30 min
- Princip testu:Quantitative
- Životnost police:Store for 6 months at 4 °C
- Detekční rozsah:31,25 - 2000 pg/ml
- Skladovací teplota:4 °C
- Objem vzorku:100 μl
- Citlivost:18,75 pg/ml
- Regulační status:RUO
Specifications
About this item
Human TGF beta Receptor I ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human TGF beta Receptor I in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 31,25 to 2000 pg/ml
- Sensitivity: 18,75 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Human TGF beta Receptor I ELISA kit (A75895) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human TGF beta Receptor I in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for TGF beta Receptor I has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the TGF beta Receptor I present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-TGF beta Receptor I Antibody, which binds the captured TGF beta Receptor I present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of TGF beta Receptor I captured in each well. The concentration of TGF beta Receptor I can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.