Order Entry
Czech Republic
ContactUsLinkComponent
Mouse STAT6 ELISA Kit
Mouse STAT6 ELISA Kit
Katalogové číslo ANTIA75869-96
Dodavatel:  ANTIBODIES.COM
New Product
Číslo CAS:  
undefined
Mouse STAT6 ELISA Kit
Katalogové číslo ANTIA75869-96
Dodavatel:  ANTIBODIES.COM
New Product
Číslo CAS:  

Specifikace

  • Bal.:
    96 Tests
  • Doba trvání testu:
    Multiple steps
  • Typ testu:
    Sandwich
  • Formát:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Cíl:
    STAT6
  • Popis:
    Mouse STAT6 ELISA kit
  • Typ vzorku:
    Serum, plasma, tissue homogenates and other biological fluids
  • Křížová reakce:
    Mouse STAT6 ELISA kit exhibits high specificity and excellent specificity for the detection of mouse STAT6. No significant cross-reactivity or interference between STAT6 and analogues was observed.
  • Detekční metoda:
    Colorimetric
  • Doba do výsledků:
    4 h 30 min
  • Princip testu:
    Quantitative
  • Životnost police:
    Store for 6 months at 4 °C
  • Detekční rozsah:
    78,125 - 5000 pg/ml
  • Skladovací teplota:
    4 °C
  • Objem vzorku:
    100 μl
  • Citlivost:
    46,875 pg/ml
  • Regulační status:
    RUO

Specifikace

O tomto produktu

Mouse STAT6 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse STAT6 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 78,125 to 5000 pg/ml
  • Sensitivity: 46,875 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse STAT6 ELISA kit (A75869) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse STAT6 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for STAT6 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the STAT6 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-STAT6 Antibody, which binds the captured STAT6 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of STAT6 captured in each well. The concentration of STAT6 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.