- Bal.:96 Tests
- Doba trvání testu:Multiple steps
- Typ testu:Sandwich
- Formát:Pre-coated
- Primary antibody reactivity:Human
- Cíl:IRE1
- Popis:Human IRE1 ELISA kit
- Typ vzorku:Serum, plasma, tissue homogenates and other biological fluids
- Křížová reakce:Human IRE1 ELISA kit exhibits high specificity and excellent specificity for the detection of human IRE1. No significant cross-reactivity or interference between IRE1 and analogues was observed.
- Detekční metoda:Colorimetric
- Doba do výsledků:4 h 30 min
- Princip testu:Quantitative
- Životnost police:Store for 6 months at 4 °C
- Detekční rozsah:62,5 - 4000 pg/ml
- Skladovací teplota:4 °C
- Objem vzorku:100 μl
- Citlivost:37,5 pg/ml
- Regulační status:RUO
Human IRE1 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IRE1 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection range: 62,5 to 4000 pg/ml
- Sensitivity: 37,5 pg/ml
- Sample type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra assay: CV <8%, Inter assay: CV <10%
Human IRE1 ELISA kit (A78041) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human IRE1 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for IRE1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IRE1 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IRE1 Antibody, which binds the captured IRE1 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IRE1 captured in each well. The concentration of IRE1 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.