Human Untagged LRRC15 Clone
Catalog # ORIGSC305914
Supplier: OriGene
New Product
CAS Number:
Specifications
- Bal.:10 µG
- Popis:Human LRRC15 untagged clone, tag free
- Vector details:pCMV6-Entry
Specifications
About this item
LRRC15 (untagged)-Human leucine rich repeat containing 15 (LRRC15), transcript variant 2.
E. coli selection: Kanamycin (25 µg/ml)
Mammalian cell selection: Neomycin
Insert size: 1700 bp