Human Tagged ORF Clone, GPR172B (SLC52A1)
Katalogové číslo ORIGRC225737L4V
Dodavatel: OriGene
New Product
Číslo CAS:
Specifikace
- Bal.:200 µl
- Popis:GPR172B (SLC52A1) Human tagged ORF clone
- Vector details:pLenti-C-mGFP-P2A-Puro
Specifikace
O tomto produktu
Lenti ORF particles, SLC52A1 (mGFP-tagged) - Human G protein-coupled receptor 172B (GPR172B), transcript variant 1, 200 µl, >10⁷ TU/ml. This clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.
The molecular sequence of this clone aligns with the gene accession number as a point of reference only. However, individual transcript sequences of the same gene can differ through naturally occurring variations (e.g. polymorphisms), each with its own valid existence.