Order Entry
Czech Republic
ContactUsLinkComponent
 
AzoDye-1-PEG10-Azide
Catalog # AATB2413
Supplier:  AAT Bioquest
New Product
CAS Number:  
AzoDye-1-PEG10-Azide
Catalog # AATB2413
Supplier:  AAT Bioquest
New Product
CAS Number:  

Specifications

  • Bal.:
    1 mg
  • Sekvence:
    2070

Specifications

About this item

 :
Storage Temperature:Freezer
 :1013.16 g/mol