CHECKIT® Lovibond® Comparator Disk, Manganese LR, 0,1 to 0,7 mg/L
Catalog # LOVI146410
Supplier: Lovibond
CAS Number:
Specifications
- Chemická metoda:PAN
- Bal.:1 KIT
- Popis:CHECKIT® comparator disk, manganese LR
- Metoda:M2522
- Parametry:Manganese LR VARIO PP
- Rozsah:0,1 - 0,7 mg/L Mn
Specifications
About this item
CHECKIT® comparator disk, manganese LR, 0,1 to 0,7 mg/L.
- Easy colour match with a continuous colour gradient
- Simply rotate the disc easily compare against a prepared water sample
- Part of a low cost solution for fast and accurate water analysis
- Low cost, easy, fast
Continuous colour Gradient: Colour discs have a continuous color gradient making it possible to achieve a direct colour match - no need to extrapolate the result between two colour standards.
Step-By-Step Instructions: Included with each colour disc, the multi-lingual instructions provide an illustrated step-by-step explanation of how to conduct the test - ensuring that even 'non-chemists' can achieve reliable and accurate measurements quickly.
Wide Variety of Parameters: Many parameters have multiple testing ranges and methods available. Additional testing parameters can be added by purchasing additional discs and reagents - utilise the same comparator housing for all of your test.
Ordering information: CHECKIT® comparator test discs are supplied as part of a complete test kit, or can be purchased individually as a replacement to expand your testing capabilities.