Scotch-Weld™ EPX Pneumatic Applicator
Katalogové číslo MMMAEPXG50P
Dodavatel: 3M
Specifikace
- Kapacita:400 ml, 490 ml
- Popis:EPX Pneumatic applicator
- Bal.:1
Specifikace
O tomto produktu
Quickly and easily metre, mix and apply two part adhesives with Scotch-Weld™ EPX pneumatic applicator.
- Pistol grip provides comfortable handling and precise control
- Measure, mix and apply
This applicator is designed to efficiently apply 3M™ Scotch-Weld™ Structural Adhesive Duo-Pak Cartridges for fast and simple applications using a pneumatic connection. The pneumatically charged system is ideal for large volume dispensing. It’s an effective system for bonding, joining, gluing, attaching, assembling, encapsulating, potting and sealing applications.