ContactUsLinkComponent
Anti-Glycophorin A Mouse Monoclonal Antibody [clone: HIR2]
Anti-Glycophorin A Mouse Monoclonal Antibody [clone: HIR2]
  ABCAAB15009-100
 :  Abcam
Anti-Glycophorin A Mouse Monoclonal Antibody [clone: HIR2]
  ABCAAB15009-100
 :  Abcam
  • Druh protilátky:
    Primární
  • Název antigenu:
    Glycophorin A
  • Symbol antigenu:
    GYPA
  • Klonovalita:
    Monoclonal
  • Klon:
    HIR2
  • Spojení:
    Unconjugated
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • Izotyp:
    IgG2b
  • Sekvence:
    1286
  • Formulář:
    Liquid
  • Synonyma antigenu:
    GPA|PAS-2, Sialoglycoprotein alpha|GYPA|Glycophorin-A|MN sialoglycoprotein
  • Skladovací pufr:
    pH: 7,4
    Preservative: 0,097% sodium azide
    Constituents: PBS
  • Skladovací teplota:
    −20...+4 °C
  • Transportní teplota:
    20
  • Imunogen:
    Synthetic Peptide within Human GYPA. The exact immunogen used to generate this antibody is proprietary information.
  • Čištění:
    Affinity purification Protein G
  • Rozměry protilátky:
    100 µg
  • Typ balení:
    Vial
  • Bal.:
    100 µG
Mouse Monoclonal Glycophorin A antibody. Suitable for Flow Cyt, IHC-P and reacts with Human samples. Immunogen corresponding to Synthetic Peptide within Human GYPA.
Recommended dilutions: Flow cytometry: 1 µg for 10⁶ Cells.
Type: Primary
Antigen: Glycophorin A
Clonality: Monoclonal
Clone: HIR2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: