ContactUsLinkComponent
Aroclor 1248 (PCB-mixture, 48% chlorinated) 100 µg/ml in cyclohexane
Aroclor 1248 (PCB-mixture, 48% chlorinated) 100 µg/ml in cyclohexane
Catalog # EHERX20124800CY
Supplier:  EHRENSTORFER
CAS Number:  
Aroclor 1248 (PCB-mixture, 48% chlorinated) 100 µg/ml in cyclohexane
Catalog # EHERX20124800CY
Supplier:  EHRENSTORFER
CAS Number:  
Specifications
  • Typ balení:
    Glass bottle
  • Bal.:
    10 mL
  • Popis:
    Aroclor 1248 (PCB-mixture, 48% chlorinated) 100 µg/ml in cyclohexane