Aroclor 1016 (PCB-mixture, 42% chlorinated) 100 µg/ml in isooctane
Katalogové číslo EHERX20101600IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:10 mL
- Popis:Aroclor 1016 (PCB-mixture, 42% chlorinated) 100 µg/ml in isooctane