Orders LinkContactUsLinkComponent
Sample Ejection Rods
Sample Ejection Rods
Supplier:   Sampling Systems
Sample Ejection Rods
Supplier:  Sampling Systems
Certificates
Sample Ejection Rods
Sample Ejection Rods
2 Options Available Below
Product Details & Documents
Sample ejection rods are designed to facilitate the removal of samples.
  • A useful tool for removing samples
  • Choose a rod longer than the sampler being used
  • Available from stock
  • Fast delivery
The sample ejection rod is a convenient tool for removing any hard-to-eject samples collected within a sampler. Simply push the rod down the sampler shaft to eject the material.
Product Family Options
Product Information
  • Sekvence
    Popis
    Materiál
    Délka
    Availability
    Price
  • 01
    Sample ejection rod
    Nerezová ocel 316L
    700 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 5 890,00 Kč
    5 890,00 Kč
    1 * 1 KS
       5 890,00 Kč
  • 02
    Sample ejection rod
    Nerezová ocel 316L
    1100 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 7 060,00 Kč
    7 060,00 Kč
    1 * 1 KS
       7 060,00 Kč