ContactUsLinkComponent
MyeloCult™ H5100 Medium
Catalog # STMC05150
CAS Number:  
undefined
MyeloCult™ H5100 Medium
Catalog # STMC05150
CAS Number:  
Specifications
  • Bal.:
    500 mL
  • Popis:
    MyeloCult™ H5100
Specifications
About this item
Medium for long-term culture of human cells.
MyeloCult™ H5100 is a culture medium for the initiation and maintenance of myeloid long-term cultures of human hematopoietic cells and stromal cell feeder layers. The serum used in this formulation has been pre-tested and selected for its ability to support long-term myelopoiesis by primitive human hematopoietic cells (e.g. in long-term culture-initiating cell (LTC-IC) assays). MyeloCult™ H5100 requires the addition of freshly prepared Hydrocortisone.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').